PIG® Formaldehyde Spill Kit in Bucket, New Pig
Supplier: New Pig
KIT501
89178-328EA 224.9 USD
89178-328
PIG® Formaldehyde Spill Kit in Bucket, New Pig
Spill Control Materials
PIG Formaldehyde Neutralizing Loose Absorbent quickly absorbs formaldehyde, formaldehyde-based fixatives, glutaraldehyde and other aldehyde solutions
Includes PPE and cleanup tools for safe, effective spill response. Screw-on, gasketed lid helps contain residual vapors after cleanup. Container with gasketed lid can be used for temporary storage of used absorbents.
No wheels. Not UN rated.
Quickly solidifies liquid formaldehyde for easy cleanup and neutralizes the harmful vapors