Dual Crown Faceshield, Honeywell Safety
Supplier: Honeywell Safety Products
FM400DCCLC
89176-996EA 144.83 USD
89176-996
Dual Crown Faceshield, Honeywell Safety
Face Shields
Dual crown face shields provides extended, wraparound protection.
  • Made from Noryl®
  • Strong, very durable and heat resistant
  • Broader deeper, 4" crown and chin protectors provide additional protection
For use in tough working conditions.