Safety Enclosures by Labconco, METTLER TOLEDO®
Supplier: Mettler Toledo
3930310 63059033 3950310
89174-754EA 10500 USD
89174-754 89204-578 89204-582 89174-752 89204-576
Safety Enclosures by Labconco, METTLER TOLEDO®
Fume Hoods and Enclosures Balances Enclosures
METTLER TOLEDO Enclosures by Labconco are specifically designed to protect the analyst during the weighing process
  • Protection during weighing
  • Clean-Sweep™ air foil
  • Maximize containment
  • Ensure safety
The patented Clean-Sweep™ air foil will maximize the containment of powders, particulates, and fumes to ensure operator safety. The ergonomic design and interior dimensions make this product ideal for micro and analytical balances, and the granite work surface will provide the greatest stability for your products.