Bottle Rack, 12-Place BOD, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W227731
89174-476CS 113.42 USD
89174-476
Bottle Rack, 12-Place BOD, Wheaton, DWK Life Sciences
Racks Bottle Racks
Manufactured from rugged and durable PVC coated wire, this rack is designed to accommodate 300mL BOD bottles
Racks can be interlocked for safe transport or storage. Each rack offers a maximum capacity of 12 bottles.