SP Bel-Art Portable Battery Powered Engraving and Labeling Tool, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F44150-0005
89171-626EA 46.52 USD
89171-626
SP Bel-Art Portable Battery Powered Engraving and Labeling Tool, Bel-Art Products, a part of SP
Pens Stylus and Scribers
Permanently mark and label your important tools and samples. Simply hold the battery-powered engraving tool like a pen, press the power button, apply light pressure and 'write' to permanently mark most materials including glass, plastic, wood, metal, bone, and stone.
  • Portable and cordless, battery-powered
  • Engraved marks are extremely resilient against wear
  • Permanently mark most materials
  • Engraving marks will not fade if exposed to chemicals, sunlight or extreme temperatures
Convenient polyethylene stand, sold separately, holds the engraving tool when not in use and has a safe place to keep extra tip. 1.5 V, operates on 2 "AA" batteries to be ordered separately.
Certifications: CE approved.
Ordering information: Includes 1 extra engraving replacement tip. , operates on 2 "AA" batteries (not included). Recommended accessory: Engraving tool stand.