Temperature Indicator Strip, Therapak®
Supplier: Therapak, LLC
40006
89171-046PK 58.76 USD
89171-046
Temperature Indicator Strip, Therapak®
Temperature Indicating Strips
Temperature Indicator Strips indicate the temperature of the specimen within minutes after the initial collection
As an integral part of the drug testing process, the temperature indicator strip confirms that the voided specimen was indeed within temperature expectations eliminating doubt at the point of collection that a specimen was subject to adulteration. Temperature readings can be viewed in both Celsius and Fahrenheit.