Urine Cytology/FISH Testing Collection and Transportation Kit, Therapak®
Supplier: Therapak, LLC
37928
89171-030CS 909.67 USD
89171-030
Urine Cytology/FISH Testing Collection and Transportation Kit, Therapak®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Kit contains the components necessary for urine cytology / FISH testing
The primary collection container is pre-filled with a urine preservative solution suitable for maintaining specimen viability after the urine content is transferred into the container for transport to the laboratory. To use, place two ounces of urine and two vials of preservative in the blue-cap container. The container is protected with a foam insert inside a transport bag and shipping box
Certifications: 95kPa Certifified.