StoneScreen™ Kidney Stone Collection Cup, Therapak®
Supplier: Therapak, LLC
75109
89170-924CS 222.69 USD
89170-924
StoneScreen™ Kidney Stone Collection Cup, Therapak®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
Designed to capture renal calculi and intended for use with patients attempting to pass stones following extracorporeal shock wave lithotripsy
The urine-straining device is a collapsible plastic strainer cup that can be used by both men and women and includes a nylon screen at the bottom of the cup for stone capture. Once the stone is captured, it can be placed in the storage space under the lid and saved for analysis.