Hanging Dual Scale Thermometer, Thermco
Supplier: Thermco
BFZ80
89168-870EA 37.34 USD
89168-870
Hanging Dual Scale Thermometer, Thermco
Thermometers Analog Thermometers
This non-mercury hanging thermometer offers a dual scale of Celsius and Fahrenheit
It works for both refrigerators and freezers, and the tube is completely enclosed in plastic for safety. Includes stainless steel shelf hangers with keyhole backs for wall hanging