Personna® Food Blades, Accutec Blades
Supplier: Accutec Blades
60-0183-0000
89168-288CS 676.75 USD
89168-288
Personna® Food Blades, Accutec Blades
Razors and Blades
Personna® Food Blades provide super sharp edges for all cutting needs
Made from stainless steel and ground to perfection, these 0.022" stainless steel Poultry blades deliver precise cuts and incredibly sharp edges for consistently clean results.
Ordering information: Blades are individually wrapped, and packaged 150 per box.