Destaining solution, Blue-OUT™ for mass spectrometry

Supplier: G-Biosciences
Danger

786-683
89167-894EA 164.96 USD
89167-894
Destaining solution, Blue-OUT™ for mass spectrometry
Destaining solution

Blue-OUT™ for the removal of Coomassie and fluorescent stain prior to mass spectrometry


Blue-OUT™ is a unique product that washes the interfering Coomassie and fluorescent protein stains from gel pieces prior to their treatment for protein sequencing or mass spectrometry.


Blue-OUT™ is also supplied in the InGel™ Blue kit for the tryptic digestion and peptide extraction of Coomassie stained proteins.

Storage Temperature: Ambient
UN: 1648
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR