Swift™ Film Cleaner, G-Biosciences
Supplier: G-Biosciences
786-678
89167-888EA 224.65 USD
89167-888
Swift™ Film Cleaner, G-Biosciences
Blotting Paper
G-Biosciences' Swift Film Cleaner™ allows researchers to clean film that has been overexposed or has a high background or speckling without having to repeat experiments.
G-Biosciences' Swift Film Cleaner™ is suitable for all experiments that use film to develop the result, including gel shift assays, Western, Southern and Northern blots. The following procedure is suitable for all developed films, new and old, and takes a few minutes to reduce the background. The cleaner is quickly stopped once the correct exposure has been reached.
Ordering information: Accessories available.