Mag-Bind® Total RNA and Total RNA 96 Kits, Omega Bio-tek®
Supplier: OMEGA BIO-TEK
M6731-01
89167-360EA 1397.75 USD
89167-360
Mag-Bind® Total RNA and Total RNA 96 Kits, Omega Bio-tek®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Mag-Bind Total® RNA Kit and Mag-Bind® Total RNA 96 Kit both provide a novel technology for total RNA isolation
These kits allow the rapid and reliable isolation of high-quality, total cellular RNA from a wide variety of cells and tissues. The system combines the higher nucleic acid-binding properties of Mag-Bind® Particles with the E.Z.N.A.® RNA Isolation system to provide a superior quality of RNA. Unlike column-based systems, the binding of nucleic acids to magnetic particles occurs in solution, resulting in increased binding kinetics and binding efficiency. Particles are also completely re-suspended during the wash steps of the purification protocol, which enhances contact with the wash buffer and removal of contaminants, thus increasing nucleic acid purity. The E.Z.N.A.® Mag-Bind® Total RNA procedure can be fully automated with most robotic workstations.