J-KEM® Dual Temperature Controllers, Chemglass
Supplier: Chemglass
CG-3210-01 CG-3210-02 CG-3210-03 CG-3210-100
89166-644EA 3557.54 USD
89166-644 89166-646 89166-648 10034-466
J-KEM® Dual Temperature Controllers, Chemglass
Controllers Temperature Controllers
J-Kem® Scientific Apollo dual temperature controller features two independent temperature controllers in a single cabinet.
  • Temperature ramping and serial communications are also provided with this unit
Each independent channel has its own high power outlet, 100 hour timer to turn heating/cooling on or off at a user set time, and solid state over temperature protection circuit. .
Ordering information: Types T,J and K are supplied complete with PTFE Thermocouple (1/8 x 12"), Coiled Extension Cord (10') and 14/20 PTFE Thermocouple Adapter. The RTD Unit is the controller only.