Rotary Evaporator Bump Traps, Elliptical, Chemglass
Supplier: Chemglass
CG-1320-E-10
89166-576EA 226.46 USD
89166-576
Rotary Evaporator Bump Traps, Elliptical, Chemglass
Vacuum Traps
Evaporator bump trap prevents contents of flask from being drawn into the condenser.
  • This allows the use of larger recovery flasks in a rotary evaporator
  • Traps have a 24/40 top outer joint and the listed size of lower inner joint
Rotary evaporator bump trap is used between the vapor tube and evaporation flask on a rotary evaporator to prevent contents of the flask from being drawn into the condenser in the event of 'bumping'. Unique elliptical shape provides large trap volume while decreasing the overall length when compared to standard spherical designs.