Hoshizaki B-300PF Ice Storage Bin
Supplier: Nordon/Hoshizaki Llc.
B-300SF
89166-528EA 2429.11 USD
89166-528
Hoshizaki B-300PF Ice Storage Bin
Ice Makers
The ice bin features a vinyl clad exterior for easy cleaning and a long lasting attractive appearance.
  • Designed with a sturdy construction for side by side or stacked installation
  • Non-corrosive bin liner provides safe sanitary storage of ice cubes or flaked ice
  • Foamed-in-place polyurethane insulation in all bin walls, and bottom, provides dependable ice storage
  • New design increases ice accessibility by 30%
HoshiGuard Antimicrobal Agents add extra protection to the storage bin and ice scoop.
Certifications: AHRI and NSF certified.
Ordering information: Ice bin only; ice maker is not included. Supplied with manufacturer's two-year warranty on parts and labor.