Fluorescein-5-maleimide ≥90% (by HPLC), Ultrapure

Supplier: Enzo Life Sciences

ENZ-52502
89166-052EA 409.84 USD
89166-052
Fluorescein-5-maleimide ≥90% (by HPLC), Ultrapure
Fluorescein-5-maleimide

Thiol dye.


Fluorescein-5-maleimide enables protein labeling via thiol modifications in most proteins at cysteine residues that either are intrinsically present or resulted from reduction of cystine residues. Unlike iodoacetamides, maleimides do not react with histidine and methionine under physiological conditions. The probe is suitable for assessing conformation- dependent accessibility of cysteine residues in proteins. Wavelength Maxima: Excitation 493 nm, Emission 515 nm.


Replaces former ENZ-52505

Formula: C₂₄H₁₃NO₇
Storage Temperature: Freezer
MDL Number: MFCD00077345
CAS Number: 75350-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR