bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥95% (by HPLC), Ultrapure dye

Supplier: Enzo Life Sciences

ENZ-52401
89166-026EA 187.48 USD
89166-026
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥95% (by HPLC), Ultrapure dye
bisbenzimide H-33342 trihydrochloride (Hoechst 33342)

DNA dye


Hoechst 33342 is fluorescent probe useful for detecting DNA by fluorescence microscopy and flow cytometry. Hoechst 33342 may be used on live or fixed cells and is also applicable for cell cycle analysis and monitoring DNA condensation by flow cytometry. An additional ethyl group renders Hoechst 33342 more lipophilic, and thus more readily cell permeable than Hoechst 33258 . Exclusion of Hoechst 33342 dye is a characteristic common to stem cells, as well as chemotherapy-resistant cancer cells. Wavelength Maxima: Excitation 350nm, Emission 461nm

Formula: C₂₇H₂₈N₆O·3HCl
Storage Temperature: Freezer
MDL Number: MFCD00012679
CAS Number: 23491-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4beb6fa5e65e2f3bb60723371acbfd3961cbdff&vid=43eadb5493e654016a2c0c73dfa9a9a8a65aaf25&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR