Dihydrorhodamine 123 ≥95% (by HPLC), Ultrapure

Supplier: Enzo Life Sciences

Synonyms: Methyl 2-(3,6-diamino-9H-xanthen-9-yl)benzoate, 2-(3,6-Diamino-9H-xanthene-9-yl)-benzoic acid methyl ester

ENZ-52302
89166-010EA 228.9 USD
89166-010
Dihydrorhodamine 123 ≥95% (by HPLC), Ultrapure
Dihydrorhodamine 123

Mitochondria dye


Dihydrorhodamine 123 is a widely utilized probe for detection of intracellular reactive oxygen species (ROS) such as peroxide, hypochlorous acid and peroxynitrite. It is readily oxidized into rhodamine 123, which exhibits a spectral profile similar to that of FITC. In combination with other fluorescent probes (such as surface receptor-targeted fluorescent antibodies, the cell viability probe propidium iodide, or fluorescent calcium indicators) dihydrorhodamine 123 can be used for multiparametric cell measurements. Wavelength Maxima: Excitation 507nm, Emission 529nm

Formula: C₂₁H₁₈N₂O₃
MW: 346.386 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04221428
CAS Number: 109244-58-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8259b2960338bd97e1060229abc58fc2d85c4973&vid=429f575ccb96e1cc5178b107d7f93eed4902ecee&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR