BCECF AM ester ≥95% (by HPLC), Ultrapure

Supplier: Enzo Life Sciences

Synonyms: 2′,7′-Bis(2-carboxyethyl)-5(6′)-carboxyfluorescein acetoxymethyl ester, Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-2',7'-dipropanoic acid, BCECF AM

ENZ-52102
89165-974EA 319.37 USD
89165-974
BCECF AM ester ≥95% (by HPLC), Ultrapure
BCECF AM ester

pH indicator


BCECF AM (2’,7’-bis-(2-carboxyethyl)-5-(and-6)-carboxyfluorescein acetoxymethyl ester) is a well established fluorescent probe that enables ratiometric monitoring of cellular pH. Intracellular pH plays an important role in modulating cellular events, including growth, calcium regulation, enzymatic activity, receptor-mediated signal transduction, ion transport, endocytosis, chemotaxis, adhesion and other cellular processes. Once inside, cellular esterases cleave the AM groups yielding a more hydrophilic BCECF trapped inside the cell. Wavelength Maxima: Excitation 505nm, Emission 520nm

Formula: C₄₂H₄₀O₂₁
MW: 808.69 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036969
CAS Number: 117464-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8c0404448381cc691798096a804b82ee29a57d2&vid=856b228fbb3a33e05bf364252e85894be3d2ac1d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR