Rhod-2 AM (Bromide) ≥90% (by HPLC), Ultrapure

Supplier: Enzo Life Sciences

ENZ-52010
89165-950EA 413.11 USD
89165-950
Rhod-2 AM (Bromide) ≥90% (by HPLC), Ultrapure
Rhod-2 AM (Bromide)

Calcium dye


Rhod-2 AM has the longest fluorescent emission signal of the commonly used calcium indicators. It contains a rhodamine-like fluorophore, making it suitable for use with argon and krypton laser excitation sources. Rhod-2 signal intensity one of the strongest of all the calcium probes. Rhod 2 is especially suitable probe for intracellular calcium monitoring by confocal laser-scanning microscopy and flow cytometry. The dissociation constant of Rhod 2 with calcium is among the highest of all the fluorescent calcium probes, providing a wider range for monitoring calcium concentration. Rhod-2 AM is an acetoxymethyl ester derivative of Rhod-2 that can be easily loaded into cells by passive incubation. Wavelength Maxima: Excitation 549nm, Emission 578nm

Formula: C₅₂H₅₉BrN₄O₁₉
Storage Temperature: Freezer
CAS Number: 145037-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df957863eba74024e5de53f43ea719cb82a53839&vid=e8d3205e12d16aaad9adbed42cd84b118122763c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR