ROS-ID® Total ROS/Superoxide Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-51010
89165-882EA 368.42 USD
89165-882
ROS-ID® Total ROS/Superoxide Detection Kit, Enzo Life Sciences
Assays Cellular Assays
The kit is designed to directly monitor real time reactive oxygen species (ROS) production and Specifically Superoxide in live cells using fluorescence microscopy or flow cytometry
Through the combination of these two specific fluorescent probes, the kit provides a simple and specific assay for the real-time measurement of global levels of reactive oxygen species (ROS), and specifically superoxide in living cells. The Total ROS/Superoxide Detection Kit contains sufficient reagents for at least 200 microscopy assays or 50 flow cytometry assays using live cells (adherent or in suspension) or 2 x 96 microplate assays.
Delivery information: Kit contains Oxidative Stress Detection Reagent (Green), 300 nmoles, Superoxide Detection Reagent (Orange), 300 nmoles, ROS Inducer (Pyocyanin), 1 µmole, ROS Inhibitor (N-acetyl-L-cysteine), 2 x 10 mg and Wash Buffer Salts, 1 pack.