Fluorescein-12-UTP, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-42834
89165-846EA
473.06
USD
89165-846
Fluorescein-12-UTP, Enzo Life Sciences
Nucleic Acid Reagents
Nucleotides
Fluorescein-12-UTP is ideal for in vitro transcription reactions catalyzed by T3, T7, or SP6 RNA polymerases and reverse transcriptase to produce fluorophore-labeled RNA transcripts. RNA-based probes can be used to identify complementary sequences by in situ hybridization to fixed cells and tissues by direct or indirect fluorescence detection or by membrane hybridization procedures.