Cyanine 3-UTP (enhanced), Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-42505
89165-754EA 462.16 USD
89165-754
Cyanine 3-UTP (enhanced), Enzo Life Sciences
Nucleic Acid Reagents Nucleotides
Cyanine 3-UTP (Enhanced) can replace UTP as a substrate for T7 RNA polymerase in labeling systems that generate labeled probes through in vitro transcription. Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP .