BIO-PROBE® BK Virus Probe, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-40848
89165-734EA 415.29 USD
89165-734
BIO-PROBE® BK Virus Probe, Enzo Life Sciences
Nucleic Acid Reagents Primers and Probes
The BK Virus BioProbe® labeled probe is prepared by nick translation of a clone of the entire BK virus genome.
BK virus, a simian polyoma virus, is related to SV40 and also bears some similarity to the human virus JC virus. BK virus is highly tumorigenic in hamsters. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).