Ubiquitin Vinyl Sulfone (Ha-Tagged), HA Tag
Supplier: Enzo Life Sciences
UW0155-0025
89165-110EA 385.86 USD
89165-110
Ubiquitin Vinyl Sulfone (Ha-Tagged), HA Tag
Proteins and Peptides
Synthetic.
For use in the detection and identification of deubiquitinylating enzymes. HA-Ub-VS is a DUB active site directed probe generated by a chemical ligation method, that acts as a potent and irreversible inhibitor of DUBs through covalent modification of the active site and as a specific probe for enzymes with DUB activity. The HA peptide sequence (YPYDVPDYA), derived from the influenza hemagglutinin protein, facilitates sensitive identification or purification of HA-Ub-VS modified DUBs through recognition by anti-HA antibodies and/or anti-HA-agarose.