Cytochalasin D (from Zygosporium mansoni) ≥98%

Supplier: Enzo Life Sciences

Synonyms: 7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-triene-1,17-dione 21(R)-acetate

BML-T109-0001
89164-970EA 141.7 USD
89164-970
Cytochalasin D (from Zygosporium mansoni) ≥98%
Cytochalasin D

Cytochalasin D is a cell-permeable fungal toxin which binds to the barbed end of actin filaments inhibiting both the association and dissociation of subunits. It causes the disruption of actin filaments and inhibition of actin polymerization. It is about 10-fold more effective than cytochalasin B and does not inhibit monosaccharide transport across cell membranes. Inactivates low conductance K+ channels. Modulates CD4 cross-linking in T-lymphocytes and increases intracellular Ca2+ levels. Exhibits antitumor activity. Induces apoptosis.


  • Ultra-pure
  • Inhibits actin polymerization
  • K+ channel blocker

Formula: C₃₀H₃₇NO₆
MDL Number: MFCD00077706
CAS Number: 22144-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66b998196c4cc5408c69d1ed6f720292d27fabdd&vid=58578e5f55072e9e9b905f9d38cd1ef3e920c7b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR