Nocodazole ≥98%

Supplier: Enzo Life Sciences

BML-T101-0010 BML-T101-0050
89164-956EA 129.71 USD
89164-956 89164-958
Nocodazole ≥98%
Nocodazole

Microtubule inhibitor


Synthetic reversible microtubule inhibitor.

Formula: C₁₄H₁₁N₃O₃S
MDL Number: MFCD00005588
CAS Number: 31430-18-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff82e70bd1808b1f01e023316722f05868369081&vid=af80ac0aecaa68226652609dbaaf2a1895b0ec35&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR