Human Recombinant Shp-2 (from E. coli)
Supplier: Enzo Life Sciences
BML-SE273-0020
89163-922EA 595.14 USD
89163-922
Human Recombinant Shp-2 (from E. coli)
Proteins and Peptides
Produced in E. coli. full length human Shp-2 (aa 1-593).