Anti-14-3-3 gamma Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA477-0100
89163-146EA 133.31 USD
89163-146 89163-148
Anti-14-3-3 gamma Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot, Affinity Chromatography

Recognizes human, mouse, rat, cow, sheep and chicken 14-3-3gamma.

Type: Primary
Antigen: 14-3-3 gamma
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR