Anti-MMP12 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA467-0100
89163-124EA 417.47 USD
89163-124
Anti-MMP12 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes human, mouse and rat MMP-12.

Type: Primary
Antigen: MMP12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR