Anti-PSMA6 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW9390-0001
89162-354EA 175.49 USD
89162-354
Anti-PSMA6 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Western Blot

Recognizes human, mouse and rabbit proteasome 20S (alpha6 subunit).

Type: Primary
Antigen: PSMA6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR