Anti-Ubiquitin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Enzo Life Sciences
PW0935-0025 PW0935-0100
89161-958EA 164.59 USD
89161-958 89161-960
Anti-Ubiquitin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Ubiquitin modification of cellular proteins has been shown to be involved with a wide range of biochemical processes including proteasomal degradation, signal transduction, DNA repair, endocytosis and autophagy.

  • For Western Blot

Recognizes mono- and poly-ubiquitin protein conjugates, free polyubiquitin chains and free ubiquitin.

Type: Primary
Antigen: UBIQUITIN
Clonality: Monoclonal
Clone:
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species Independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR