Anti-Proteasome 19S Rpn12/S14 Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW0440-0025
89161-840EA 241.98 USD
89161-840
Anti-Proteasome 19S Rpn12/S14 Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes Arabidopsis proteasome 26S (19S regulator complex) (Rpn12a/S14 subunit).

Type: Primary
Antigen: PROTEASOME 19S RPN12/S14 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Plant (Arabidopsis Thaliana)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR