Anti-Neurofilament Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
NA1223-0100
89160-646EA 301.93 USD
89160-646
Anti-Neurofilament Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat and other mammalian species neurofilament triplet proteins.

Type: Primary
Antigen: NEUROFILAMENT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR