Sodium dodecyl sulfate (SDS)

Supplier: Enzo Life Sciences
Danger

BML-KI341-0012
89160-374EA 49.05 USD
89160-374
Sodium dodecyl sulfate (SDS)
Sodium dodecyl sulphate (SDS)

Component of the 20S proteasome drug discovery assay.

Formula: CH₃(CH₂)₁₁OSO₃Na
Storage Temperature: Refrigerator
MDL Number: MFCD00036175
CAS Number: 151-21-3
UN: 2926
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR