Minoxidil ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Warning

Synonyms: 2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide, Rogaine, Loniten

BML-KC125-0500
89160-204EA 439.27 USD
89160-204
Minoxidil ≥98% (by HPLC)
Minoxidil

K⁺ channel activator.


  • Formulation: White solid.


A selective ATP-sensitive K⁺ (KATP) channel opener. It is metabolised to the more active sulphate in vivo. A clinically useful anti-alopecia. Antihypertensive. Stimulates renin secretion. Potent vascular smooth muscle relaxant.


Soluble in 100% ethanol (29 mg/mL), DMSO (6.5 mg/mL) or water (2.2 mg/mL).

Formula: C₉H₁₅N₅O
MW: 209.253 g/mol
MDL Number: MFCD00063409
CAS Number: 38304-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b5385668cecc96cfa6b146cafb35271670aad84&vid=0487d1352f3e327bf97a9d568dcb276d9b2189af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR