Actinomycin D (from Streptomyces parvulus) ≥98% (by TLC)

Supplier: Enzo Life Sciences
Danger

BML-GR300-0005
89158-858EA 99.19 USD
89158-858
Actinomycin D (from Streptomyces parvulus) ≥98% (by TLC)
Actinomycin D

An intercalating transcription inhibitor and antineoplastic antibiotic


Inhibits transcription by binding to DNA at dG residues. Induces apoptosis in many cell lines, but suppresses in PC12 cells induced by topoisomrease II inhibitor etopside. Used at 5-10 µg/mL in cell culture for halting transcription. The product is not sterile.


Red-orange crystalline solid


Soluble in DMSO (>20 mg/mL), 100% ethanol (1 mg/mL) or methanol

Formula: C₆₂H₈₆N₁₂O₁₆
Storage Temperature: Freezer
MDL Number: MFCD00005033
CAS Number: 50-76-0
UN: 3462
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cde9b7d28f0132d28e65c4135b94a831303eb34&vid=d38d0403aca86c9ce2901f80c447277fbdf95b76&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR