Isotetrandrine ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-G520-0001
89158-648EA 94.83 USD
89158-648
Isotetrandrine ≥98% (by TLC)
Isotetrandrine

Phospholipase inhibitor


Isotetrandrine is a biscoclaurine alkaloid which inhibits G-protein mediated activation of phospholipase A2 (PLA2) but not phospholipase C (PLC) or D (PLD). Cell permeable.

Formula: C₃₈H₄₂N₂O₆
MW: 622.762 g/mol
MDL Number: MFCD00236404
CAS Number: 477-57-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05c178dedb6bb0a4a20ee2cb0f0fbbcfcda89c9e&vid=af0d3ebf00527a8e8e8e1317e09e0f239cd677c4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR