Geldanamycin ≥98% (by TLC) (conjugated to FITC)

Supplier: Enzo Life Sciences

BML-EI361-0001
89158-178EA 350.98 USD
89158-178
Geldanamycin ≥98% (by TLC) (conjugated to FITC)
Geldanamycin

A novel geldanamycin fluorescent probe that may be used in a fluorescence polarization assay for HSP90 inhibitors. May also be used for detection of cell surface HSP90.


  • HSP90 inhibitor

Formula: C₂₉H₄₀N₂O₉
Storage Temperature: Freezer
MDL Number: MFCD00274570
CAS Number: 30562-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9f2eeda2fade38200833570477fa8aea3b9351b&vid=ca3bc4812d992c8dc72883d4d186a936443924df&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR