QNZ ≥98%

Supplier: Enzo Life Sciences

Synonyms: 6-Amino-4-[2-(4-phenoxyphenyl)ethylamino]quinazoline, N4-(4-Phenoxyphenethyl)quinazoline-4,6-diamine

BML-EI352-0001
89158-158EA 179.85 USD
89158-158
QNZ ≥98%
QNZ

NF-kappaB inhibitor


Inhibitor of NF-κB which displays potent inhibitory effects on both NF-κB transcriptional activation (IC50=11nM) and TNF-α production (IC50= 7nM). A useful tool to explore the role of NF-κB in cellular signaling pathways.

Formula: C₂₂H₂₀N₄O
MW: 356.429 g/mol
MDL Number: MFCD06411436
CAS Number: 545380-34-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=715bc58577d0a427f85ed0fdb820378a1342e963&vid=7e9741c3b4bbdfb2415c909687739354d76a7fdc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR