Geldanamycin ≥98% Biotin

Supplier: Enzo Life Sciences

BML-EI341-0001
89158-122EA 305.2 USD
89158-122
Geldanamycin ≥98% Biotin
Geldanamycin

A useful tool for affinity purification of HSP-90 and HSP-90 dependent client proteins.


  • HSP90 inhibitor

Formula: C₂₉H₄₀N₂O₉
MDL Number: MFCD00274570
CAS Number: 30562-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32b45bb0e5ddbf857980143d9e7b85cb1d2fa765&vid=6f0f481f8e03338983d73cf2d19294c43448e9b3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR