Wedelolactone ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 1,8,9-Trihydroxy-3-methoxy-6H-benzofuro[3,2-c]chromen-6-one, 7-Methoxy-5,11,12-trihydroxy-coumestan

BML-EI316-0001
89158-054EA 158.05 USD
89158-054
Wedelolactone ≥98% (by TLC)
Wedelolactone

IKK inhibitor


Wedelolactone is a naturally occuring furanocoumarin isolated from Eclipta prostrata, an herbal medicine used to treat septic shock in traditional Chinese medicine. It inhibits the IκB kinase (IKK) complex and supresses LPS-induced caspase-11 expression in LPS-treated BALB 3T3 cells (IC50=35µM)

Formula: C₁₆H₁₀O₇
MDL Number: MFCD07778564
CAS Number: 524-12-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13284235d273f77eb9c7e9e61b5e35b54107dd19&vid=bf781b3dda68d24821fe6261fddb3e9a42c338dc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR