H-89 dihydrochloride ≥99% (by TLC)

Supplier: Enzo Life Sciences

BML-EI196-0005 BML-EI196-0025
89157-758EA 163.5 USD
89157-758 89157-760
H-89 dihydrochloride ≥99% (by TLC)
H-89 dihydrochloride

Protein kinase inhibitor


Cell permeable potent and selective inhibitor of cAMP- and cGMP-dependent protein kinases (PKA, Ki=48nM, and PKG) and protein kinase Cµ (PKCµ). In contrast, most other protein kinase C (PKC) isotypes are much more weakly inhibited. Also inhibits Ca2+/calmodulin-dependent protein kinase II, casein kinase I and myosin light chain kinase. Induces apoptosis. Concentration range for cultured cells 30-100μM: osteoblastic cells, renal proximal tubule cells and Xenopus oocytes. 

Formula: C₂₀H₂₀BrN₃O₂S·2HCl
CAS Number: 127243-85-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c653d8d9cbba21852182d8a40af0436506ed4642&vid=96dbf46c2fd5bfb81f62d481cab999c131245ff9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR