Phloretin ≥98%

Supplier: Enzo Life Sciences

BML-EI154-0200
89157-668EA 88.29 USD
89157-668 89157-670
Phloretin ≥98%
Phloretin

PKC inhibitor


Flavonoid. Protein kinase C (PKC) inhibitor. PGF2a receptor antagonist. In astrocytes, these receptors are linked to phospholipase C, thus affecting phosphoinositide hydrolysis and intracellular Ca2+ levels. Inhibits myo-inositol uptake and 5’-iodothyronine deiodinase. Prevents TNF-α stimulated upregulation of VCAM-1, ICAM-1 and E-selectin. Enhances adiponectin expression.

Formula: C₁₅H₁₄O₅
MW: 274.28 g/mol
MDL Number: MFCD00002288
CAS Number: 60-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ff90a03f79b14180e9effab5feccb72b2b294dd&vid=437fe8e03845f5ecd1e62f70d14e001def50ee3f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR