1-O-Hexadecyl-2-O-acetyl-sn-glycerol ≥95% (by TLC)

Supplier: Enzo Life Sciences

BML-EI150-0005
89157-658EA 163.5 USD
89157-658
1-O-Hexadecyl-2-O-acetyl-sn-glycerol ≥95% (by TLC)
1-O-Hexadecyl-2-O-acetyl-sn-glycerol

Inhibits protein kinase C activation by diacylglycerols. Stimulates HL-60 cell differentiation. Biosynthetic precursor of PAF via remodeling and retroconversion/de novo routes. Induces delayed aggregation of rabbit platelets. Mitogenic for smooth muscle cells.


  • PKC inhibitor


Waxy solid.


Soluble in 100% ethanol or acetone.

Formula: C₂₁H₄₂O₄
MW: 358.6 g/mol
Storage Temperature: Freezer
CAS Number: 77133-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e96ca7441b0289668df555fb61a4ed353c68db86&vid=92ef5550fd7080fede2cf15dc76244733b168a12&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR