Erbstatin analog ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: Erbstatin Analog, 2,5-Dihydroxycinnamic acid methyll ester, Methyl 2,5-Dihydroxycinnamate, Methyl 3-(2,5-dihydroxyphenyl)acrylate

BML-EI146-0005
89157-650EA 127.53 USD
89157-650
Erbstatin analog ≥98% (by TLC)
Erbstatin analog

EGFR inhibitor


Erbstatin is a novel inhibitor of the EGF receptor-associated tyrosine kinase. It is an unstable compound and is completely inactivated in serum in 30 minutes. Methyl 2,5-dihydroxycinnamate is a stable erbstatin analog retaining activity after a 60 minute incubation. It inhibits EGF receptor-associated tyrosine kinase in vitro (IC50=0.77 µM). Inhibition is competitive with substrate and noncompetitive with ATP.

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
MDL Number: MFCD00132932
CAS Number: 63177-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bcc0e44de7da485af66b11a6d64b316f77f1430&vid=ca9f04ff404d98389fffa94dd3980c4a49f2bf49&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR