Baicalein ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one

BML-EI106-0050
89157-568EA 104 USD
89157-568 89157-570
Baicalein ≥98% (by TLC)
Baicalein

Lipoxygenase inhibitor


Inhibits cellular Ca2+ uptake and calcium mobilization. Inhibitor of protein tyrosine kinase in leukemia (CEM) cells. Induces cell cycle arrest and apoptosis. Anti-inflammatory compound. Has anti-thrombotic, anti-proliferative and anti-mitogenic effects.Originally reported as a selective inhibitor of 12-lipoxygenase, baicalein has since been shown to inhibit both 12-LO and 15-LO (IC50=0.64µM for 12-LO and 1.6µM for 15-LO). Inhibits leukotriene biosynthesis (at higher concentrations) and release of lysosomal enzymes. Inhibits adjuvant-induced arthritis. Also inhibits prolyl oligopeptidase (POP/PREP)(IC50=36µM).

Formula: C₁₅H₁₀O₅
MDL Number: MFCD00017459
CAS Number: 491-67-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b20c55630ac1375c425f570fc4fed1bfc99e885&vid=c959da995a9d69eacfbe28e3244bd37454493182&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR