1-Stearoyl-2-arachidonoyl-sn-glycerol ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-DG130-0010
89157-358EA 348.8 USD
89157-358
1-Stearoyl-2-arachidonoyl-sn-glycerol ≥98% (by TLC)
1-Stearoyl-2-arachidonoyl-sn-glycerol

PKC activator.


  • Formulation: Oil or waxy solid.


Putative endogenous activator of protein kinase C produced via receptor-mediated inositol phospholipid hydrolysis.


Soluble in DMSO or 100% ethanol.

Formula: C₄₁H₇₂O₅
MW: 645.0 g/mol
Storage Temperature: Freezer
CAS Number: 65914-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9184b232c29bd1f7753afdfc3e1fd7de38eaeac8&vid=49e6e77452204d9f0e5604958dbceb4427fd0df1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥95% (TLC)
Appearance Oil or waxy solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR