BAPTA-AM ≥90% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: Tetrakis(acetoxymethyl)-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate, Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N,N-tetraacetate, 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid Tetrakis(acetoxymethyl) Ester, BAPTA-AM ester

BML-CA411-0025
89156-852EA 281.22 USD
89156-852
BAPTA-AM ≥90% (by TLC)
BAPTA-AM

PKC activator


BAPTA is a Ca2+ chelator with 105-fold greater affinity for Ca2+ than for Mg2+. BAPTA-AM is a cell-permeable derivative which is widely used as an intracellular calcium sponge. Induces inactivation of protein kinase C. Also inhibits thapsigargin-induced apoptosis in rat thymocytes.

Formula: C₃₄H₄₀N₂O₁₈
MW: 764.69 g/mol
MDL Number: MFCD00036696
CAS Number: 126150-97-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24411035ec098b3798a373e5f9598dbfa1cad807&vid=fbe0308895aaa94ad89ae9adecba938098511b27&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR